3-Bromo-2-fluoro-5-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36609-500MG 36609-1G 36609-5G 36609-25G
77326-162EA 218.36 USD
77326-162 77349-868 77362-486 77385-394
3-Bromo-2-fluoro-5-(methoxymethyl)benzaldehyde ≥97%
3-Bromo-2-fluoro-5-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632046
CAS Number: 1891083-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR