Methyl 4-isopropoxy-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36599-500MG 36599-1G 36599-5G
77335-386EA 353.29 USD
77335-386 77349-862 77374-072
Methyl 4-isopropoxy-3-(trifluoromethyl)benzoate ≥97%
Methyl 4-isopropoxy-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O₃
MW: 262.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632043
CAS Number: 1401511-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR