1-Bromo-2-(methoxymethyl)-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

36596-1G 36596-5G 36596-25G
77340-648EA 60.77 USD
77340-648 77362-482 77374-070
1-Bromo-2-(methoxymethyl)-4-(trifluoromethyl)benzene ≥95%
1-Bromo-2-(methoxymethyl)-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O
MW: 269.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632042
CAS Number: 1646161-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR