4-(Benzyloxy)-1-bromo-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36591-500MG 36591-1G 36591-5G 36591-25G
77326-156EA 103 USD
77326-156 77343-030 77362-480 77374-068
4-(Benzyloxy)-1-bromo-2-(methoxymethyl)benzene ≥97%
4-(Benzyloxy)-1-bromo-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
MW: 307.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632039
CAS Number: 2379322-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR