1-Chloro-2,4-difluoro-5-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

36590-500MG 36590-1G 36590-5G 36590-25G
77326-154EA 233.81 USD
77326-154 77349-860 77362-478 77385-390
1-Chloro-2,4-difluoro-5-(methoxymethyl)benzene ≥95%
1-Chloro-2,4-difluoro-5-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD32632038
CAS Number: 2379322-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR