1-Bromo-2,4-difluoro-5-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

36588-500MG 36588-1G 36588-5G 36588-25G
77326-152EA 243.08 USD
77326-152 77349-858 77362-476 77385-388
1-Bromo-2,4-difluoro-5-(methoxymethyl)benzene ≥95%
1-Bromo-2,4-difluoro-5-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD32632037
CAS Number: 2379322-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR