1,3-Dichloro-5-fluoro-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36587-500MG 36587-1G 36587-5G 36587-25G
77326-150EA 243.08 USD
77326-150 77349-856 77362-474 77385-386
1,3-Dichloro-5-fluoro-2-(methoxymethyl)benzene ≥97%
1,3-Dichloro-5-fluoro-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD32632036
CAS Number: 2379322-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR