(3-Fluoro-2-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

36585-500MG 36585-1G 36585-5G 36585-25G
77326-148EA 105.06 USD
77326-148 77343-072 77362-472 77374-066
(3-Fluoro-2-isopropoxyphenyl)methanol ≥97%
(3-Fluoro-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26056743
CAS Number: 1565737-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR