tert-Butyl 3-fluoro-5-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

36577-500MG 36577-1G 36577-5G 36577-25G
77326-146EA 218.36 USD
77326-146 77349-854 77362-470 77385-384
tert-Butyl 3-fluoro-5-(methylthio)benzoate ≥95%
tert-Butyl 3-fluoro-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₂S
MW: 242.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632032
CAS Number: 2379322-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR