Ethyl 3-fluoro-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

36576-500MG 36576-1G 36576-5G 36576-25G
77326-144EA 204.97 USD
77326-144 77349-852 77362-468 77385-382
Ethyl 3-fluoro-5-(methylthio)benzoate ≥97%
Ethyl 3-fluoro-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34178498
CAS Number: 2643368-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR