tert-Butyl 3-bromo-2-fluoro-5-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

36572-500MG 36572-1G 36572-5G
77337-490EA 415.09 USD
77337-490 77349-850 77374-064
tert-Butyl 3-bromo-2-fluoro-5-(trifluoromethyl)benzoate ≥95%
tert-Butyl 3-bromo-2-fluoro-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrF₄O₂
MW: 343.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32265228
CAS Number: 2379322-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR