Ethyl 4-(benzyloxy)-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

36570-500MG 36570-1G 36570-5G 36570-25G
77326-142EA 193.64 USD
77326-142 77349-848 77362-466 77385-380
Ethyl 4-(benzyloxy)-3-fluorobenzoate ≥97%
Ethyl 4-(benzyloxy)-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₃
MW: 274.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632031
CAS Number: 1451154-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR