Methyl 4-(benzyloxy)-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

36569-500MG 36569-1G 36569-5G 36569-25G
77326-140EA 90.64 USD
77326-140 77342-382 77362-464 77374-062
Methyl 4-(benzyloxy)-3-fluorobenzoate ≥97%
Methyl 4-(benzyloxy)-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632030
CAS Number: 2379322-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR