1-(Benzyloxy)-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36566-1G 36566-5G 36566-25G
77341-454EA 87.55 USD
77341-454 77362-462 77374-060
1-(Benzyloxy)-3-isopropoxybenzene ≥97%
1-(Benzyloxy)-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₂
MW: 242.313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632029
CAS Number: 585509-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR