1-(Benzyloxy)-3-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36565-1G 36565-5G 36565-25G
77341-398EA 84.46 USD
77341-398 77362-460 77374-058
1-(Benzyloxy)-3-ethoxybenzene ≥97%
1-(Benzyloxy)-3-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632028
CAS Number: 2296808-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR