(2-Ethoxynaphthalen-1-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

36559-500MG 36559-1G 36559-5G 36559-25G
77326-136EA 181.28 USD
77326-136 77346-822 77362-454 77385-376
(2-Ethoxynaphthalen-1-yl)methanol ≥97%
(2-Ethoxynaphthalen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₂
MW: 202.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03715925
CAS Number: 690963-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR