1,2-Dibromo-3,5-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36543-1G 36543-5G 36543-25G
77341-994EA 111.24 USD
77341-994 77362-450 77374-050
1,2-Dibromo-3,5-dimethoxybenzene ≥95%
1,2-Dibromo-3,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD23827949
CAS Number: 191847-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR