3-Chloro-2-(2-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

36522-500MG 36522-1G 36522-5G
77335-384EA 353.29 USD
77335-384 77349-840 77374-046
3-Chloro-2-(2-fluorophenyl)pyridine ≥97%
3-Chloro-2-(2-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClFN
MW: 207.631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698922
CAS Number: 387827-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR