3-Chloro-2-(4-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

36520-500MG 36520-1G 36520-5G
77335-382EA 353.29 USD
77335-382 77349-838 77374-044
3-Chloro-2-(4-fluorophenyl)pyridine ≥97%
3-Chloro-2-(4-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClFN
MW: 207.631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698924
CAS Number: 847226-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR