3-Chloro-2-(3-methoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

36518-500MG 36518-1G 36518-5G
77335-380EA 353.29 USD
77335-380 77349-836 77374-042
3-Chloro-2-(3-methoxyphenyl)pyridine ≥97%
3-Chloro-2-(3-methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD30098129
CAS Number: 847225-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR