2,5-Dimethoxy-4-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36503-500MG 36503-1G 36503-5G
77339-584EA 602.55 USD
77339-584 77349-830 77374-038
2,5-Dimethoxy-4-(methoxymethyl)benzoic acid ≥97%
2,5-Dimethoxy-4-(methoxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD32632019
CAS Number: 1898237-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR