1-Ethoxy-2,3,4-trifluorobenzene ≥97%

Supplier: AOBChem USA

Warning

36486-1G 36486-5G 36486-25G
77341-094EA 71.07 USD
77341-094 77362-446 77374-036
1-Ethoxy-2,3,4-trifluorobenzene ≥97%
1-Ethoxy-2,3,4-trifluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇F₃O
MW: 176.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29970689
CAS Number: 1862370-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR