4-Isopropoxy-1,2-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36483-1G 36483-5G 36483-25G
77341-092EA 71.07 USD
77341-092 77362-444 77374-034
4-Isopropoxy-1,2-dimethoxybenzene ≥97%
4-Isopropoxy-1,2-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD23113855
CAS Number: 77106-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR