4-Ethoxy-1,2-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36482-1G 36482-5G 36482-25G
77340-646EA 60.77 USD
77340-646 77362-442 77374-032
4-Ethoxy-1,2-dimethoxybenzene ≥97%
4-Ethoxy-1,2-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00673003
CAS Number: 41827-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR