(2-Fluoro-5-(methoxymethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

36481-500MG 36481-1G 36481-25G 36481-5G
77326-128EA 292.52 USD
77326-128 77349-828 77385-370 77374-030
(2-Fluoro-5-(methoxymethyl)phenyl)methanol ≥97%
(2-Fluoro-5-(methoxymethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632018
CAS Number: 1895357-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR