Methyl 2-fluoro-5-(methoxymethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36479-500MG 36479-1G 36479-5G
77336-782EA 394.49 USD
77336-782 77349-824 77374-028
Methyl 2-fluoro-5-(methoxymethyl)benzoate ≥97%
Methyl 2-fluoro-5-(methoxymethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632016
CAS Number: 1870852-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR