1-(Benzyloxy)-5-bromo-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36425-500MG 36425-1G 36425-5G 36425-25G
77326-120EA 141.11 USD
77326-120 77344-794 77362-432 77385-364
1-(Benzyloxy)-5-bromo-2-fluoro-3-methylbenzene ≥97%
1-(Benzyloxy)-5-bromo-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632013
CAS Number: 2404734-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR