1-Bromo-4-isopropoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

36421-1G 36421-5G 36421-25G
77340-950EA 64.89 USD
77340-950 77362-430 77374-022
1-Bromo-4-isopropoxynaphthalene ≥97%
1-Bromo-4-isopropoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD32632009
CAS Number: 185310-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR