tert-Butyl 2,6-dichloro-4-(trifluoromethyl)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
36418-500MG
36418-1G
36418-5G
36418-25G
77326-118EA
243.08
USD
77326-118
77349-818
77362-428
77385-362
tert-Butyl 2,6-dichloro-4-(trifluoromethyl)benzoate ≥97%
tert-Butyl 2,6-dichloro-4-(trifluoromethyl)benzoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁Cl₂F₃O₂ MW: 315.116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32632007 CAS Number: 2379322-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...