Ethyl 2,6-dichloro-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36417-500MG 36417-1G 36417-5G 36417-25G
77326-116EA 218.36 USD
77326-116 77349-816 77362-426 77385-360
Ethyl 2,6-dichloro-4-(trifluoromethyl)benzoate ≥97%
Ethyl 2,6-dichloro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32632006
CAS Number: 189338-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR