Methyl 2,6-dichloro-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36416-500MG 36416-1G 36416-5G 36416-25G
77326-114EA 218.36 USD
77326-114 77349-814 77362-424 77385-358
Methyl 2,6-dichloro-4-(trifluoromethyl)benzoate ≥97%
Methyl 2,6-dichloro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32632005
CAS Number: 2379322-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR