2-(benzo[d]thiazol-2-yl)-4-methylphenol ≥95%

Supplier: AOBChem USA

Warning

36413-500MG 36413-1G 36413-5G
77326-112EA 340.93 USD
77326-112 77349-812 77374-018
2-(benzo[d]thiazol-2-yl)-4-methylphenol ≥95%
2-(benzo[d]thiazol-2-yl)-4-methylphenol

Precaución: for Research Use Only

Formula: C₁₄H₁₁NOS
MW: 241.308 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11180245
CAS Number: 21703-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR