1-(3-Bromo-4-methylphenyl)-1H-pyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

36406-500MG 36406-1G 36406-5G 36406-25G
77326-110EA 193.64 USD
77326-110 77349-810 77362-422 77385-356
1-(3-Bromo-4-methylphenyl)-1H-pyrazole-4-carbaldehyde ≥97%
1-(3-Bromo-4-methylphenyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O
MW: 265.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27989933
CAS Number: 1780427-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR