1-(3-Bromo-4-methylphenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

36404-500MG 36404-1G 36404-5G
77336-132EA 378.01 USD
77336-132 77349-808 77374-016
1-(3-Bromo-4-methylphenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥95%
1-(3-Bromo-4-methylphenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrN₂O
MW: 293.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11959835
CAS Number: 1175817-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR