1-(3-Bromo-4-methylphenyl)-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

36403-500MG 36403-1G 36403-5G 36403-25G
77326-108EA 119.48 USD
77326-108 77343-632 77362-420 77374-014
1-(3-Bromo-4-methylphenyl)-3,5-dimethyl-1H-pyrazole ≥97%
1-(3-Bromo-4-methylphenyl)-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂
MW: 265.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11926463
CAS Number: 1176493-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR