1-(3-Bromo-4-methylphenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

36402-500MG 36402-1G 36402-5G 36402-25G
77326-106EA 119.48 USD
77326-106 77343-630 77362-418 77374-012
1-(3-Bromo-4-methylphenyl)-1H-pyrazole ≥97%
1-(3-Bromo-4-methylphenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD27989931
CAS Number: 1823931-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR