2-(1H-benzo[d]imidazol-2-yl)-4-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

36393-500MG 36393-1G 36393-5G 36393-25G
77326-104EA 232.78 USD
77326-104 77349-806 77362-416 77385-354
2-(1H-benzo[d]imidazol-2-yl)-4-fluorophenol ≥97%
2-(1H-benzo[d]imidazol-2-yl)-4-fluorophenol

Precaución: for Research Use Only

Formula: C₁₃H₉FN₂O
MW: 228.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23629770
CAS Number: 112291-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR