Methyl 5-ethoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

36392-500MG 36392-1G 36392-5G
77326-102EA 317.24 USD
77326-102 77349-804 77374-010
Methyl 5-ethoxynicotinate ≥97%
Methyl 5-ethoxynicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27931050
CAS Number: 874285-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR