1,3-Dibromo-5-fluoro-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36383-500MG 36383-1G 36383-5G
77338-306EA 473.8 USD
77338-306 77349-802 77374-004
1,3-Dibromo-5-fluoro-2-(methoxymethyl)benzene ≥97%
1,3-Dibromo-5-fluoro-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD32632003
CAS Number: 2379322-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR