5,7-Dibromo-3-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

36381-500MG 36381-1G 36381-25G 36381-5G
77326-098EA 292.52 USD
77326-098 77349-800 77385-352 77374-002
5,7-Dibromo-3-iodo-1H-indazole ≥97%
5,7-Dibromo-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₃Br₂IN₂
MW: 401.825 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13177075
CAS Number: 1131632-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR