1,2-Bis(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

36376-1G 36376-5G 36376-25G
77341-224EA 77.25 USD
77341-224 77362-410 77374-000
1,2-Bis(methoxymethoxy)benzene ≥95%
1,2-Bis(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MW: 198.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28133434
CAS Number: 3688-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR