1-Ethoxy-3,4-difluoro-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36350-500MG 36350-1G 36350-5G
77326-092EA 332.69 USD
77326-092 77349-794 77373-992
1-Ethoxy-3,4-difluoro-2-methoxybenzene ≥95%
1-Ethoxy-3,4-difluoro-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32632000
CAS Number: 2379321-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR