1,2-Difluoro-3,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36349-500MG 36349-1G 36349-5G
77326-090EA 341.96 USD
77326-090 77349-792 77373-990
1,2-Difluoro-3,4-dimethoxybenzene ≥97%
1,2-Difluoro-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28788789
CAS Number: 172657-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR