2-Benzyloxy-5-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-(Benzyloxy)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Benzyloxy-5-fluorobenzeneboronic acid pinacol ester

36329-1G 36329-5G 36329-25G
77341-128EA 72.1 USD
77341-128 77362-402 77373-982
2-Benzyloxy-5-fluorophenylboronic acid pinacol ester ≥97%
2-Benzyloxy-5-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD22494088
CAS Number: 779331-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR