(2-Methoxy-5-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

36323-500MG 36323-1G 36323-5G
77335-678EA 363.59 USD
77335-678 77349-788 77373-980
(2-Methoxy-5-(methylthio)phenyl)methanol ≥97%
(2-Methoxy-5-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
MW: 184.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29927610
CAS Number: 904311-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR