2-Bromo-4-fluoro-1-methoxy-3-(methoxymethyl)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
36322-500MG
36322-1G
36322-5G
36322-25G
77326-088EA
193.64
USD
77326-088
77349-786
77362-400
77385-350
2-Bromo-4-fluoro-1-methoxy-3-(methoxymethyl)benzene ≥95%
2-Bromo-4-fluoro-1-methoxy-3-(methoxymethyl)benzene
Precaución: for Research Use Only
Formula:
C₉H₁₀BrFO₂ MW: 249.077 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32631997 CAS Number: 2379321-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...