2-Bromo-5-(methoxymethyl)-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

36248-500MG 36248-1G 36248-5G
77339-886EA 671.56 USD
77339-886 77349-776 77373-976
2-Bromo-5-(methoxymethyl)-3-methylpyridine ≥97%
2-Bromo-5-(methoxymethyl)-3-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631988
CAS Number: 2379321-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR