1-Bromo-2-fluoro-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36245-500MG 36245-1G 36245-5G 36245-25G
77326-078EA 202.91 USD
77326-078 77349-774 77362-392 77385-340
1-Bromo-2-fluoro-3-(methoxymethyl)benzene ≥97%
1-Bromo-2-fluoro-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28347685
CAS Number: 1307255-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR