1,3-Difluoro-2-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

36237-1G 36237-5G 36237-25G
77340-644EA 60.77 USD
77340-644 77362-388 77373-974
1,3-Difluoro-2-(methoxymethyl)benzene ≥95%
1,3-Difluoro-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631981
CAS Number: 110213-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR