(3-Bromo-2-butoxy-6-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

36226-500MG 36226-1G 36226-5G 36226-25G
77326-072EA 218.36 USD
77326-072 77349-770 77362-386 77385-334
(3-Bromo-2-butoxy-6-fluorophenyl)methanol ≥95%
(3-Bromo-2-butoxy-6-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD32631980
CAS Number: 2379321-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR